Bond Elut adapters
Nome del fornitore: VARIAN
Vai a
Informazioni su questo articolo
Bond Elut adapters fit on top of any Bond Elut cartridge and contain a female Luer fitting that accommodates the tip of another cartridge.
- Connect SPE cartridges in series for large samples
- Expand cartridge volume for even more applications
- Transfer large-volume samples to any SPE cartridge
2 varianti
Product Details & Documents
Bond Elut adapters fit on top of any Bond Elut cartridge and contain a female Luer fitting that accommodates the tip of another cartridge.
- Connect SPE cartridges in series for large samples
- Expand cartridge volume for even more applications
- Transfer large-volume samples to any SPE cartridge
Product Family Options
Informazioni sul prodotto
- DisponibilitàPrice
- VARI12131003Adapter cap for 12 and 20 ml Bond Elut cartridges
- VARI12131004Adapter cap for 60 ml Bond Elut cartridges
This product has been blocked by your organization. Please contact your purchasing department for more information