Orders LinkContactUsLinkComponent
Graphite tubes for PerkinElmer
Graphite tubes for PerkinElmer
Nome del fornitore:   AGILENT HEWLETT PACKARD
Graphite tubes for PerkinElmer
Nome del fornitore:  AGILENT HEWLETT PACKARD
Certificates
Graphite tubes for PerkinElmer
Graphite tubes for PerkinElmer
7 varianti
Product Details & Documents
These graphite platforms and tubes for PerkinElmer graphite furnace AA systems are quality tested to ensure every batch passes our demanding performance specifications for contamination, sensitivity, precision, electrical resistance, and lifetime.
  • Guaranteed to be compatible with PerkinElmer instruments
  • Backed by the stand behind warranty that guarantees the parts will not cause instrument failure or downtime
  • Parts qualified on a genuine PerkinElmer system
Use partitioned tubes for general-purpose work and for the best detection limits with samples that have low background. These are also ideal when measuring organic solutions, or samples with low viscosity that may spread throughout the tube, potentially degrading precision.
Instruction guides and how-to videos available to ensure you get the best performance from your PerkinElmer system when using our parts and supplies.
Product Family Options
Informazioni sul prodotto
  • Per
    Disponibilità
    Price
  • HEWL8003-0962
    Graphite tube, pyrocoated, HGA, not compatible with L'vov platforms
    Graphite tube, pyrocoated, HGA, not compatible with L'vov platforms
    HGA 900/850/800/700/600/300 furnaces
    Log in per disponibilità
    Log in per disponibilità
    1 * 10 ST
    1 * 10 ST
  • HEWL8003-0954
    Graphite tube, pyrocoated, HGA, with advanced platform
    Graphite tube, pyrocoated, HGA, with advanced platform
    HGA 900/850/800/700/600/300 furnaces
    Log in per disponibilità
    Log in per disponibilità
    1 * 5 ST
    1 * 5 ST
  • HEWL8003-0416
    Graphite tube, pyrocoated, HGA, with integrated spherical platform that accommodates up to 50 ml sample
    Graphite tube, pyrocoated, HGA, with integrated spherical platform that accommodates up to 50 ml sample
    Volatile and nonvolatile refractory elements, for HGA 300/600/700/800/850/900 furnaces
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST
  • HEWL8003-0421
    Graphite tube, pyrocoated, with grooves for L'vov platforms (not including platform)
    Graphite tube, pyrocoated, with grooves for L'vov platforms (not including platform)
    HGA 300/600/700/800/850/900 furnaces
    Log in per disponibilità
    Log in per disponibilità
    1 * 10 ST
    1 * 10 ST
  • HEWL8003-0959
    Graphite tube, standard, THGA
    Graphite tube, standard, THGA
    THGA graphite furnace systems
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST
  • HEWL8003-0958
    Graphite tube, standard, THGA, with end cap
    Graphite tube, standard, THGA, with end cap
    THGA graphite furnace systems
    Log in per disponibilità
    Log in per disponibilità
    1 * 5 ST
    1 * 5 ST
  • HEWL8003-0410
    Graphite tube, standard, THGA, with integrated platform, for THGA graphite furnace systems
    Graphite tube, standard, THGA, with integrated platform, for THGA graphite furnace systems
    THGA graphite furnace systems
    Log in per disponibilità
    Log in per disponibilità
    1 * 5 ST
    1 * 5 ST