Tuning and performance standards for LC/MS
Tuning solution kit, 500 LC/MS, 1
Codice catalogo VARI393210201
Nome del fornitore: VARIAN
Tuning and performance standards for LC/MS
Codice catalogo VARI393210201
Nome del fornitore: VARIAN
Numero CAS:
Vai a
Product Details & Documents
Agilent offers a range of tuning and performance standards for LC/MS.
- Typical standards have at least a year of shelf-life
- Many standards align with a range of applications
- Unique formulations that are tested and verified on Agilent instrumentation
A critical part of developing a solution, keeping your instrument qualified, and even trusting the instrument’s results is to regularly test the efficacy of the instrument. Agilent offers a range of certified chemical standards that are integral to instrument, performance, and application verification for your LC and LC/MS system. You can rest assured that Agilent tuning mixes are of the highest quality –designed, developed, and manufactured to guarantee at least one year of shelf-life.
Informazioni per l'ordine: Most standards manufactured with ISO 17025 and Guide 34 certification.
Confezione: Environmentally friendly packaging that is easy-to-open and verify information.
Specifiche
Specifiche
- Conf:1
- Descrizione:Tuning solution kit, 500 LC/MS