Square Buckets
Nome del fornitore: LAMAPLAST
Vai a
Informazioni su questo articolo
Robust PP bucket with ergonomic handle.
- Square shape
- Flat bottom ensures total emptying and easy cleaning
- Food contact material approved
Hot filling possible up to 95 °C.
2 varianti
Product Details & Documents
Robust PP bucket with ergonomic handle.
- Square shape
- Flat bottom ensures total emptying and easy cleaning
- Food contact material approved
Hot filling possible up to 95 °C.