Orders LinkContactUsLinkComponent
Matrix™ Tri-Coded Tubes
Matrix™ Tri-Coded Tubes
   Opzioni sostenibili disponibili
Nome del fornitore:   THERMO MATRIX TECHNOLOGIES
Matrix™ Tri-Coded Tubes
Nome del fornitore:  THERMO MATRIX TECHNOLOGIES
   Opzioni sostenibili disponibili
Certificates
Matrix™ Tri-Coded Tubes
19 varianti
Product Details & Documents
Matrix™ tri-coded tubes combine the 2D matrix code with an improved linear and human-readable barcode.
  • The linear and human-readable codes match the tubes’ unique 2D code to allow 1D scanning or visual identification of samples across collaborative sites without 2D readers
  • An expanded offering of tri-coded tubes to provide more flexibility for your work
  • The high-contrast barcodes improve the reliability of reading and identifying samples
  • Enhanced sample security with three different barcode formats: 1D (side), human readable (side), 2D barcode (bottom)
Enhance sample tracking with Matrix Tri-coded Storage Tubes and avoid taking chances when storing and sharing precious samples. With the rapid growth of biobanking, infectious disease research and development, cancer research and molecular diagnostics, storing valuable samples is more important than it has ever been within the scientific community.
Matrix™ storage tubes feature permanent high-contrast 2D, 1D linear, and human-readable barcodes, offering three levels of identification to safely track valuable samples. Matrix storage tubes come in sterile and non-sterile options and can withstand many years of freezer storage and repeated freeze/thaw cycles.
Matrix storage tubes enable:
• Elimination of secondary labeling steps—2D-barcoded Matrix™ ScrewTop and Open-Top Storage Tubes are available preprinted with 1D and human-readable codes that match the tubes’ 2D barcodes.
• Enhanced secure tracking—Permanent 2D barcode is laser-etched onto the base of every tube to securely identify and track samples. The linear and human-readable barcodes match this unique 2D barcode, allowing 1D scanning or visual identification of samples across satellite sites without 2D readers.
• Stringent quality control—Every 2D barcoded storage tube is scanned to ensure readability and uniqueness.
Specifications for Product Family
Specifiche
  • Catalog No:
  • 479-2453
  • 479-2450
  • 479-1050
  • 479-2454
  • 479-1049
  • 479-2455
  • 479-2456
  • 479-1055
  • 479-2457
  • 479-2458
  • 479-1035
  • 479-2459
  • 479-1034
  • 479-2449
  • 479-1039
  • 479-2460
  • 479-2461
  • 479-2462
  • 479-2463
  • Environmentally Preferable:
  • Produzione a basso impatto
  • Produzione a basso impatto
  • Produzione a basso impatto
  • Produzione a basso impatto
  • Produzione a basso impatto
  • Produzione a basso impatto
  • Produzione a basso impatto
  • Produzione a basso impatto
  • Produzione a basso impatto
  • Produzione a basso impatto
  • Produzione a basso impatto
  • Produzione a basso impatto
  • Produzione a basso impatto
  • Produzione a basso impatto
  • Produzione a basso impatto
  • Produzione a basso impatto
  • Produzione a basso impatto
  • Produzione a basso impatto
  • Produzione a basso impatto
  • Certificazione ambientale:
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
Specifiche
Product Family Options
0,5 ml Tubes (max. working volume 450 µl)
Informazioni sul prodotto
  • Capacità
    Fondo
    Tappo incluso
    Colore tappo
    Sterili
    Confezione
    Disponibilità
    Price
  • 479-2453
    2D white patch 1D linear barcode, barcoded rack
    2D white patch 1D linear barcode, barcoded rack
    0,50 ml
    V bottom
    Yes
    Colour-21390374|Colour-21390348
    -
    5 racks of 96/case
    Log in per disponibilità
    Log in per disponibilità
    1 * 480 ST
    1 * 480 ST - SFr. 781.00
    SFr. 781.00
    1 * 480 ST
       SFr. 781.00
    Specifiche:
      Produzione a basso impatto
    More
    Specifications
    Cat. No.
    479-2453
    Environmentally Preferable
    Produzione a basso impatto
    Certificazione ambientale
    NAR Certificate of Retirement
      Produzione a basso impatto
  • 479-2450
    2D white patch 1D linear barcode, barcoded rack
    2D white patch 1D linear barcode, barcoded rack
    0,50 ml
    V bottom
    Yes
    Colour-21390374|Colour-21390348
    +
    5 racks of 96/case
    Log in per disponibilità
    Log in per disponibilità
    1 * 480 ST
    1 * 480 ST - SFr. 924.00
    SFr. 924.00
    1 * 480 ST
       SFr. 924.00
    Specifiche:
      Produzione a basso impatto
    More
    Specifications
    Cat. No.
    479-2450
    Environmentally Preferable
    Produzione a basso impatto
    Certificazione ambientale
    NAR Certificate of Retirement
      Produzione a basso impatto
  • 479-1050
    2D white patch 1D linear barcode
    2D white patch 1D linear barcode
    0,50 ml
    V bottom
    Yes
    Colour-21390374|Colour-21390348
    +
    5 racks of 96/case
    Log in per disponibilità
    Log in per disponibilità
    1 * 480 ST
    1 * 480 ST - SFr. 712.00
    SFr. 712.00
    1 * 480 ST
       SFr. 712.00
    Specifiche:
      Produzione a basso impatto
    More
    Specifications
    Cat. No.
    479-1050
    Environmentally Preferable
    Produzione a basso impatto
    Certificazione ambientale
    NAR Certificate of Retirement
      Produzione a basso impatto
  • 479-2454
    2D white patch, barcoded rack
    2D white patch, barcoded rack
    0,50 ml
    V bottom
    Yes
    Colour-21390374|Colour-21390348
    +
    5 racks of 96/case
    Log in per disponibilità
    Log in per disponibilità
    1 * 480 ST
    1 * 480 ST - SFr. 720.00
    SFr. 720.00
    1 * 480 ST
       SFr. 720.00
    Specifiche:
      Produzione a basso impatto
    More
    Specifications
    Cat. No.
    479-2454
    Environmentally Preferable
    Produzione a basso impatto
    Certificazione ambientale
    NAR Certificate of Retirement
      Produzione a basso impatto
  • 479-1049
    2D white patch
    2D white patch
    0,50 ml
    V bottom
    Yes
    Colour-21390374|Colour-21390348
    +
    5 racks of 96/case
    Log in per disponibilità
    Log in per disponibilità
    1 * 480 ST
    1 * 480 ST - SFr. 655.00
    SFr. 655.00
    1 * 480 ST
       SFr. 655.00
    Specifiche:
      Produzione a basso impatto
    More
    Specifications
    Cat. No.
    479-1049
    Environmentally Preferable
    Produzione a basso impatto
    Certificazione ambientale
    NAR Certificate of Retirement
      Produzione a basso impatto
  • 479-2455
    2D white patch 1D linear barcode, barcoded rack
    2D white patch 1D linear barcode, barcoded rack
    0,50 ml
    V bottom
    No
    -
    5 racks of 96/case
    Log in per disponibilità
    Log in per disponibilità
    1 * 480 ST
    1 * 480 ST - SFr. 503.00
    SFr. 503.00
    1 * 480 ST
       SFr. 503.00
    Specifiche:
      Produzione a basso impatto
    More
    Specifications
    Cat. No.
    479-2455
    Environmentally Preferable
    Produzione a basso impatto
    Certificazione ambientale
    NAR Certificate of Retirement
      Produzione a basso impatto
  • 479-2456
    2D white patch 1D linear barcode, barcoded rack
    2D white patch 1D linear barcode, barcoded rack
    0,50 ml
    V bottom
    No
    +
    5 racks of 96/case
    Log in per disponibilità
    Log in per disponibilità
    1 * 480 ST
    1 * 480 ST - SFr. 546.00
    SFr. 546.00
    1 * 480 ST
       SFr. 546.00
    Specifiche:
      Produzione a basso impatto
    More
    Specifications
    Cat. No.
    479-2456
    Environmentally Preferable
    Produzione a basso impatto
    Certificazione ambientale
    NAR Certificate of Retirement
      Produzione a basso impatto
  • 479-1055
    2D white patch 1D linear barcode
    2D white patch 1D linear barcode
    0,50 ml
    V bottom
    No
    +
    5 racks of 96/case
    Log in per disponibilità
    Log in per disponibilità
    1 * 480 ST
    1 * 480 ST - SFr. 499.00
    SFr. 499.00
    1 * 480 ST
       SFr. 499.00
    Specifiche:
      Produzione a basso impatto
    More
    Specifications
    Cat. No.
    479-1055
    Environmentally Preferable
    Produzione a basso impatto
    Certificazione ambientale
    NAR Certificate of Retirement
      Produzione a basso impatto
1,0 ml Tubes (max. working volume 925 µl)
Informazioni sul prodotto
  • Capacità
    Fondo
    Tappo incluso
    Colore tappo
    Sterili
    Confezione
    Disponibilità
    Price
  • 479-2457
    2D white patch 1D linear barcode
    2D white patch 1D linear barcode
    1,00 ml
    V bottom
    Yes
    Colour-21390374|Colour-21390348
    +
    Bulk, 480/case
    Log in per disponibilità
    Log in per disponibilità
    1 * 480 ST
    1 * 480 ST - SFr. 567.00
    SFr. 567.00
    1 * 480 ST
       SFr. 567.00
    Specifiche:
      Produzione a basso impatto
    More
    Specifications
    Cat. No.
    479-2457
    Environmentally Preferable
    Produzione a basso impatto
    Certificazione ambientale
    NAR Certificate of Retirement
      Produzione a basso impatto
  • 479-2458
    2D white patch 1D linear barcode, barcoded rack
    2D white patch 1D linear barcode, barcoded rack
    1,00 ml
    V bottom
    Yes
    Colour-21390374|Colour-21390348
    +
    5 racks of 96/case
    Log in per disponibilità
    Log in per disponibilità
    1 * 480 ST
    1 * 480 ST - SFr. 760.00
    SFr. 760.00
    1 * 480 ST
       SFr. 760.00
    Specifiche:
      Produzione a basso impatto
    More
    Specifications
    Cat. No.
    479-2458
    Environmentally Preferable
    Produzione a basso impatto
    Certificazione ambientale
    NAR Certificate of Retirement
      Produzione a basso impatto
  • 479-1035
    2D white patch 1D linear barcode
    2D white patch 1D linear barcode
    1,00 ml
    V bottom
    Yes
    Colour-21390374|Colour-21390348
    +
    5 racks of 96/case
    Log in per disponibilità
    Log in per disponibilità
    1 * 480 ST
    1 * 480 ST - SFr. 712.00
    SFr. 712.00
    1 * 480 ST
       SFr. 712.00
    Specifiche:
      Produzione a basso impatto
    More
    Specifications
    Cat. No.
    479-1035
    Environmentally Preferable
    Produzione a basso impatto
    Certificazione ambientale
    NAR Certificate of Retirement
      Produzione a basso impatto
  • 479-2459
    2D white patch, barcoded rack
    2D white patch, barcoded rack
    1,00 ml
    V bottom
    Yes
    Colour-21390374|Colour-21390348
    +
    5 racks of 96/case
    Log in per disponibilità
    Log in per disponibilità
    1 * 480 ST
    1 * 480 ST - SFr. 738.00
    SFr. 738.00
    1 * 480 ST
       SFr. 738.00
    Specifiche:
      Produzione a basso impatto
    More
    Specifications
    Cat. No.
    479-2459
    Environmentally Preferable
    Produzione a basso impatto
    Certificazione ambientale
    NAR Certificate of Retirement
      Produzione a basso impatto
  • 479-1034
    2D white patch
    2D white patch
    1,00 ml
    V bottom
    Yes
    Colour-21390374|Colour-21390348
    +
    5 racks of 96/case
    Log in per disponibilità
    Log in per disponibilità
    1 * 480 ST
    1 * 480 ST - SFr. 655.00
    SFr. 655.00
    1 * 480 ST
       SFr. 655.00
    Specifiche:
      Produzione a basso impatto
    More
    Specifications
    Cat. No.
    479-1034
    Environmentally Preferable
    Produzione a basso impatto
    Certificazione ambientale
    NAR Certificate of Retirement
      Produzione a basso impatto
  • 479-2449
    2D white patch 1D linear barcode, barcoded rack
    2D white patch 1D linear barcode, barcoded rack
    1,00 ml
    V bottom
    No
    +
    5 racks of 96/case
    Log in per disponibilità
    Log in per disponibilità
    1 * 480 ST
    1 * 480 ST - SFr. 546.00
    SFr. 546.00
    1 * 480 ST
       SFr. 546.00
    Specifiche:
      Produzione a basso impatto
    More
    Specifications
    Cat. No.
    479-2449
    Environmentally Preferable
    Produzione a basso impatto
    Certificazione ambientale
    NAR Certificate of Retirement
      Produzione a basso impatto
  • 479-1039
    2D white patch 1D linear barcode
    2D white patch 1D linear barcode
    1,00 ml
    V bottom
    No
    +
    5 racks of 96/case
    Log in per disponibilità
    Log in per disponibilità
    1 * 480 ST
    1 * 480 ST - SFr. 499.00
    SFr. 499.00
    1 * 480 ST
       SFr. 499.00
    Specifiche:
      Produzione a basso impatto
    More
    Specifications
    Cat. No.
    479-1039
    Environmentally Preferable
    Produzione a basso impatto
    Certificazione ambientale
    NAR Certificate of Retirement
      Produzione a basso impatto
  • 479-2460
    2D white patch, barcoded rack
    2D white patch, barcoded rack
    1,00 ml
    V bottom
    No
    +
    5 racks of 96/case
    Log in per disponibilità
    Log in per disponibilità
    1 * 480 ST
    1 * 480 ST - SFr. 480.00
    SFr. 480.00
    1 * 480 ST
       SFr. 480.00
    Specifiche:
      Produzione a basso impatto
    More
    Specifications
    Cat. No.
    479-2460
    Environmentally Preferable
    Produzione a basso impatto
    Certificazione ambientale
    NAR Certificate of Retirement
      Produzione a basso impatto
1,4 ml Open-top tubes (max. working volume 1260 µl)
Informazioni sul prodotto
  • Capacità
    Fondo
    Tappo incluso
    Colore tappo
    Sterili
    Confezione
    Disponibilità
    Price
  • 479-2461
    2D white patch 1D linear barcode
    2D white patch 1D linear barcode
    1,40 ml
    V bottom
    No
    -
    Bulk, 1000/case
    Log in per disponibilità
    Log in per disponibilità
    1 * 1.000 ST
    1 * 1.000 ST - SFr. 927.00
    SFr. 927.00
    1 * 1.000 ST
       SFr. 927.00
    Specifiche:
      Produzione a basso impatto
    More
    Specifications
    Cat. No.
    479-2461
    Environmentally Preferable
    Produzione a basso impatto
    Certificazione ambientale
    NAR Certificate of Retirement
      Produzione a basso impatto
  • 479-2462
    2D white patch 1D linear barcode, barcoded rack
    2D white patch 1D linear barcode, barcoded rack
    1,40 ml
    V bottom
    No
    -
    10 latch racks of 96 tubes/case
    Log in per disponibilità
    Log in per disponibilità
    1 * 480 ST
    1 * 480 ST - SFr. 1’070.00
    SFr. 1’070.00
    1 * 480 ST
       SFr. 1’070.00
    Specifiche:
      Produzione a basso impatto
    More
    Specifications
    Cat. No.
    479-2462
    Environmentally Preferable
    Produzione a basso impatto
    Certificazione ambientale
    NAR Certificate of Retirement
      Produzione a basso impatto
  • 479-2463
    2D white patch 1D linear barcode, barcoded rack
    2D white patch 1D linear barcode, barcoded rack
    1,40 ml
    V bottom
    No
    +
    10 latch racks of 96 tubes/case
    Log in per disponibilità
    Log in per disponibilità
    1 * 480 ST
    1 * 480 ST - SFr. 1’060.00
    SFr. 1’060.00
    1 * 480 ST
       SFr. 1’060.00
    Specifiche:
      Produzione a basso impatto
    More
    Specifications
    Cat. No.
    479-2463
    Environmentally Preferable
    Produzione a basso impatto
    Certificazione ambientale
    NAR Certificate of Retirement
      Produzione a basso impatto
Accessories
Informazioni sul prodotto
  • Codice catalogo
    Colore
    Materiale
    Disponibilità
    Price
  • SepraSeal®, pre-split, 10 mats of 96 caps
    SepraSeal®, pre-split, 10 mats of 96 caps
    479-1247
    naturale
    Thermoplastic elastomer
    Log in per disponibilità
    1 * 960 ST - SFr. 147.00
Informazioni sul prodotto
  • Codice catalogo
    Colore
    Materiale
    Disponibilità
    Price
  • SepraSeal®, 10 mats of 96 caps
    SepraSeal®, 10 mats of 96 caps
    MATR4463
    naturale
    Thermoplastic elastomer
    Log in per disponibilità
    1 * 960 ST - SFr. 123.00
Informazioni sul prodotto
  • Codice catalogo
    Colore
    Materiale
    Disponibilità
    Price
  • SepraSeal®, 10 mats of 96 caps
    SepraSeal®, 10 mats of 96 caps
    479-1245
    naturale
    Thermoplastic elastomer
    Log in per disponibilità
    1 * 960 ST - SFr. 136.00