ContactUsLinkComponent
Excel wipes, Oxivir®
Catalog # DIVY100984247
Supplier:  Diversey
   Più sicuro dal punto di vista della salute umana e dell'ambiente
   Imballi sostenibili
undefined
Excel wipes, Oxivir®
Catalog # DIVY100984247
Supplier:  Diversey
   Più sicuro dal punto di vista della salute umana e dell'ambiente
   Imballi sostenibili
Specifications
  • Confezione:
    100/pack
  • Descrizione:
    Oxivir® Excel wipes
  • Seq:
    03
  • Environmentally Preferable:
    Più sicuro dal punto di vista della salute umana e dell'ambiente,
    Imballi sostenibili
  • Certificazione ambientale:
    ISO 1491:2007(E), including review of ISO 10993
  • Conf:
    12
Specifications
About this item
Powered by AHP® technology. Oxivir® disinfectant cleaners provide an alternative to traditional disinfectants by delivering fast acting, broad spectrum disinfection with enhanced cleaning power that is staff and environmentally friendly.
  • Broad efficacy
  • Short contact times keep the surface wet, thus ensuring disinfectant efficacy
  • Excellent cleaning performance
  • Positive user and surface safety profile
  • Fragrance-free
  • Active breaks down into water and oxygen
  • No rinsing required
Oxivir® Excel wipe CE is a one-step cleaner disinfectant wipe with broad spectrum efficacy and short contact times. Based on accelerated hydrogen peroxide formulation, these wipes can be used on non invasive medical devices.
Extensively tested against European norms and effective against multiple pathogens of concern, carries a full virucidal claim including Poliovirus, Adenovirus, and Murine norovirus, considered effective against all enveloped viruses such as: HIV, HBV, HCV, Influenza, Vaccinia virus and Coronavirus.
Certifications: Efficacy proven by compliance with EN 13727, EN 13624, EN 14476, and EN 16615. The surfactants used in this product are in compliance with the requirement of EU Detergent Regulation, EC 648/2004.
Caution: Do not mix with other products. Do not use on water- or acid-sensitive surfaces.
*I prodotti in questa pagina non rientrano nell'ambito della Direttiva UE 2019/904