Autoclavable muff covers, with drawstring, Tyvek®
Informazioni su questo articolo
Save time and standardise activities in equipment preparation for sterilisation.
- Manufactured and packed in a controlled cleanroom environment
- Reduces labour costs
- Simplifies the conformance to wrapping SOP's
- Conveniently packed: 100 per sealed poly bag
- Autoclavable at 121 to 125 °C
- Tear and puncture resistant
- Hydrophobic
- Lint-free
Tyvek® muffs provide a convenient means to cover flasks, beakers, containers, media bottles, valves, pipes and hoses. These closures are breathable and can be used to prevent particulates and microbial contamination from entering openings of any size. Since they are autoclavable, they can be used during sterilisation to reduce time-consuming wrapping of open containers. Each closure incorporates a Tyvek® drawstring closure to create the tightest possible seal around all openings.
Delivery information: Certificates of conformance sent with each shipment.
5 varianti
Product Details & Documents
Save time and standardise activities in equipment preparation for sterilisation.
- Manufactured and packed in a controlled cleanroom environment
- Reduces labour costs
- Simplifies the conformance to wrapping SOP's
- Conveniently packed: 100 per sealed poly bag
- Autoclavable at 121 to 125 °C
- Tear and puncture resistant
- Hydrophobic
- Lint-free
Tyvek® muffs provide a convenient means to cover flasks, beakers, containers, media bottles, valves, pipes and hoses. These closures are breathable and can be used to prevent particulates and microbial contamination from entering openings of any size. Since they are autoclavable, they can be used during sterilisation to reduce time-consuming wrapping of open containers. Each closure incorporates a Tyvek® drawstring closure to create the tightest possible seal around all openings.
Delivery information: Certificates of conformance sent with each shipment.
Documents
Tyvek 1422A Muffs with Drawstring - MFT2AxD
Scheda dati
Product Family Options
Informazioni sul prodotto
- ColoreDisponibilitàPrice
- 129-1769Tyvek® 1422A muff cover with drawstring, 5" (127 mm), fits 1,75 - 2,5" (44,5 - 63,5 mm) ØTyvek® 1422A muff cover with drawstring, 5" (127 mm), fits 1,75 - 2,5" (44,5 - 63,5 mm) ØbiancoLog in per disponibilità1 * 1.000 STSFr. 5’500.00
- 129-1770Tyvek® 1422A muff cover with drawstring, 7" (178 mm), fits 2,5 - 4" (63 - 102 mm) ØTyvek® 1422A muff cover with drawstring, 7" (178 mm), fits 2,5 - 4" (63 - 102 mm) ØbiancoLog in per disponibilità1 * 1.000 STSFr. 5’840.00
- 129-1771Tyvek® 1422A muff cover with drawstring, 9" (229 mm), fits 4 - 5,5" (102 - 140 mm) ØTyvek® 1422A muff cover with drawstring, 9" (229 mm), fits 4 - 5,5" (102 - 140 mm) ØbiancoLog in per disponibilità1 * 1.000 STSFr. 6’600.00
- 129-1772Tyvek® 1422A muff cover with drawstring, 12" (305 mm), fits 5,5 - 8" (140 - 203 mm) ØTyvek® 1422A muff cover with drawstring, 12" (305 mm), fits 5,5 - 8" (140 - 203 mm) ØbiancoLog in per disponibilità1 * 1.000 STSFr. 8’800.00
- 129-1773Tyvek® 1422A muff cover with drawstring, 16" (406 mm), fits 8 - 12" (203 - 305 mm) ØTyvek® 1422A muff cover with drawstring, 16" (406 mm), fits 8 - 12" (203 - 305 mm) ØbiancoLog in per disponibilità1 * 500 STSFr. 5’280.00