Distributor for capillaries, b.safe
Capillary connector, 3× capillary connector, 3× fittings for tubing Ext.Ø 3,2 mm, UNF 1/4"
Codice catalogo 590-0195
Nome del fornitore: Bohlender
Vai a
Product Details & Documents
Distributor for connection of capillary tubing. With PFA Fittings for capillaries with Ext.Ø 3,2 mm (thread UNF ¹/₄").
- Excellent chemical resistance
- Available for a wide range of hose diameters
- The conical shape gets the connection tight and leak-free
Adaptor to enlarge the number of capillary connectors on b.safe Caps.
Specifiche
- Filettatura:UNF 1/4"
- con:3× capillary connector, 3× fittings for tubing Ext.Ø 3,2 mm
- Descrizione:Capillary connector