Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
4-(4-Cyanophenyl)morpholine
4-(4-Cyanophenyl)morpholine 97%
4-(4-Cyanophenyl)morpholine 97%
APOSOR59955-1G
:
Apollo Scientific
:
4-(4-Cyanophenyl)morpholine 97%
APOSOR59955-1G
:
Apollo Scientific
:
Tipo di conf.:
Bottle
Conf:
1 g
Seq:
3047
:
:
188.23 g/mol
:
85 °C
:
GHS07
:
Ambiente
:
10282-31-2
:
C₁₁H₁₂N₂O
:
MFCD00100115