Anti-S protein RBD neutralising antibody HEK293 monoclonal antibody
Catalog # ACRBS1N-M122-1MG
Supplier: ACROBIOSYSTEMS
Specifications
- Tipo anticorpo:Primary
- Tipo di anticorpi:S protein RBD
- Simbolo dell'antigene:S protein RBD
- Clonalità:Monoclonale
- Coniugazione:Non coniugato
- ELISA:Yes
- Ospite:HEK293
- Assorbimento incrociato:This product is a specific antibody against SARS-CoV-2 Spike protein RBD domain. No cross-reactivity is deteced with Spike protein RBD domain of other coronaviruses, including SARS-CoV, MERS-CoV, HCoV-229E, HCoV-NL63, HCoV-OC43 and HCoV-HKU1.
- Forma:Powder
- Sinomini antigene:Spike,S protein RBD,Spike glycoprotein Receptor-binding domain,S glycoprotein RBD,Spike protein RBD,COVID-19
- Peso molecolare:49.2 kDa and 23.5 kDa
- Temperatura di conservazione:For long term storage, the product should be stored at lyophilised state at –20 °C or lower.
- Temperatura di spedizione:RT
- Purificazione:Affinity purified
- Conf:1 mg
Specifications
About this item
Type: Primary
Antigen: S protein RBD
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: HEK293
Isotype: Human IgG1/kappa
Reactivity: