Order Entry
Switzerland
ContactUsLinkComponent
Anti-PRPH Mouse Monoclonal Antibody [clone: 7C5]
Anti-PRPH Mouse Monoclonal Antibody [clone: 7C5]
  BSENM-1400-100
 :  Biosensis
Anti-PRPH Mouse Monoclonal Antibody [clone: 7C5]
  BSENM-1400-100
 :  Biosensis

 

  • Tipo anticorpo:
    Primary
  • Tipo di anticorpi:
    Peripherin
  • Simbolo dell'antigene:
    PRPH
  • Clonalità:
    Monoclonale
  • Clone:
    Clone: 7C5
  • Coniugazione:
    Non coniugato
  • Ospite:
    Mouse
  • ImmunoChimica:
    Yes
  • ImmunoFluorescenza:
    Yes
  • Isotipo:
    IgG
  • Western blot:
    Yes
  • Forma:
    Lyophilized
  • ID gene:
    Uniprot# P21807
  • Sinomini antigene:
    Prph1
  • Temperatura di spedizione:
    RT
  • Immunogeno:
    Recombinant full length rat Peripherin protein expressed in and purified from E.coli
  • Purificazione:
    Affinity purified
  • Conf:
    100 µG

 

 

Peripherin is a class-III neuronal intermediate filament protein found in certain classes of neuron, most of which are located in the peripheral nervous system.

Application information: Western Blotting (WB), Immunocytochemistry (IC) and Immunohistochemistry (IH). A concentration of 0.5 - 2 ug/mL is recommended for WB. A concentration of 1-5 ug/mL is recommended for IC and IH. This antibody performs well on aldehyde fixed tissues. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

"Type:
Antigen:
Clonality:
Clone:
Conjugation:
Epitope:
Host:
Isotype:
Reactivity: "