Cleanroom hoods, Tyvek® IsoClean®, model IC668B 00, non sterile
Catalog # DUPOIC668BWH00
Supplier: DuPont
Specifications
- Taglia dell'abbigliamento:One size
- Applicazioni per abiti:Cleanroom
- Genere:Unisex
- Colore:bianco
- Descrizione:Hood with ties, non-sterile
- Conf:100
- Tessuto:Tyvek®
- Resistenza tessile:>30 N
Specifications
About this item
This hood is not clean-processed and not sterilised made of Tyvek®, a flash spun high density polyethylene. The fabric is permeable to both air and water vapour, yet repels water-based liquids and aerosols. It offers an excellent barrier against fine particles and fibres (down to 1 µm in size) and is ultra-low linting.
- Full face opening with loops and ties
- Bound seams: tightly sewn seam is covered with garment fabric to reinforce seam protection and reduce potential for liquid and particle penetration
- Partial body chemical protective clothing
- Barrier to infective agents
Suitable for use in GMP class C/D (ISO Class 6-9) clean rooms. Ideal for use in the biotech, pharmaceutical, medical device manufacturing and electronics industries, as well as in other critical or controlled environments.
Certifications: Certified according to Regulation (EU) 2016/425; Partial body chemical protective clothing, Category III, Type PB [6-B]; EN 14126 (barrier to infective agents).
Packaging: Packed in cardboard box, 100 per box, bulk packed. 2 polyethylene liners.