Cleanroom overalls, Tyvek® IsoClean® model IC193B DS
: DuPont
These hooded overalls are made of Tyvek®, a fabric composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort. Tyvek® is permeable to both air and water vapour, yet repels water-based liquids and aerosols. It offers an excellent barrier against fine particles and fibres (down to 1 µm in size).
- Hooded coverall with integrated overboots
- Ultra-low linting fabric
- Tyvek® covered elasticated thumb loops
- Tunnelled elastication at wrists and ankles; stitched-in elasticated waist
- Front zipper closure with storm flap
- Optimised shape and fit allowing full freedom of movement
- Clean-processed and gamma-sterilised
- Gamma irradiated to Sterility Assurance Level (SAL) of 10⁻⁶
Garments made of Tyvek® IsoClean are used in the biotech, pharmaceutical, medical device manufacturing and electronics industries, as well as in other critical or controlled environments.
Colour: white
: Double bagged. Packed in an ISO Class 4 certified cleanroom .
- Catalog No:
- DUPOIC193BWHDS/4XL
- DUPOIC193BWHDS/7XL
- DUPOIC193BWHDS/M
- DUPOIC193BWHDS/6XL
- DUPOIC193BWHDS/S
- DUPOIC193BWHDS/2XL
- DUPOIC193BWHDS/L
- DUPOIC193BWHDS/5XL
- DUPOIC193BWHDS/XL
- DUPOIC193BWHDS/XS
- DUPOIC193BWHDS/3XL
- Modello:
- IC 193 B
- IC 193 B
- IC 193 B
- IC 193 B
- IC 193 B
- IC 193 B
- IC 193 B
- IC 193 B
- IC 193 B
- IC 193 B
- IC 193 B
- Applicazioni per abiti:
- Chemical protection, Cleanroom, Medical, Laboratory, Clinical
- Chemical protection, Cleanroom, Medical, Laboratory, Clinical
- Chemical protection, Cleanroom, Medical, Laboratory, Clinical
- Chemical protection, Cleanroom, Medical, Laboratory, Clinical
- Chemical protection, Cleanroom, Medical, Laboratory, Clinical
- Chemical protection, Cleanroom, Medical, Laboratory, Clinical
- Chemical protection, Cleanroom, Medical, Laboratory, Clinical
- Chemical protection, Cleanroom, Medical, Laboratory, Clinical
- Chemical protection, Cleanroom, Medical, Laboratory, Clinical
- Chemical protection, Cleanroom, Medical, Laboratory, Clinical
- Chemical protection, Cleanroom, Medical, Laboratory, Clinical
Frequently Bought Together