Alcian Blue, pyridine variant ≥85 % (dye content), Sigma-Aldrich®
Catalog # 335630-25G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Conf:25 g
- Seq:3308
Specifications
About this item
:
:1086.36 g/mol
Storage Temperature:Ambiente
CAS Number:123439-83-8
Formula:C₅₆H₄Cl₄CuN₁₂