Schaeffer and Fulton Spore Stain Solution A, Sigma-Aldrich®
Catalog # 90903-100ML-F
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Conf:100 mL
- Seq:2440
Specifications
About this item
:
ADR:6.1,III
:GHS08
:GHS05
:GHS09
CAS Number:2437-29-8
MDL Number:MFCD06412418