ContactUsLinkComponent
Antimicrobial extra foam hand cleanser, OxyBAC®
Antimicrobial extra foam hand cleanser, OxyBAC®
Catalog # EVONOXYEX1LBG
Supplier:  Deb-STOKO
Antimicrobial extra foam hand cleanser, OxyBAC®
Catalog # EVONOXYEX1LBG
Supplier:  Deb-STOKO
Specifications
  • Descrizione:
    Extra foam hand cleanser with 1 L cartridge
  • Conf:
    6
Specifications
About this item
Antimicrobial rich-cream foam hand wash with OptiDose™ pump, perfume-free and dye-free.
  • Contains accelerated hydrogen peroxide
  • Skin hypoallergenic
  • Suitable for food facilities and healthcare workers
  • Excellent skin cleansing
  • Hygienic cartridge
Dispenses foam on the hands which is instantly ready for washing and quick & easy rinsing from hands and sinks. Helps to prevent drying and leave the skin feeling smooth after use.
Certifications: HACCP Certified. Passes EN1499 just with ONE dose.
Caution: Use biocides safely. Always read the label and product information before use.