Aqueous mounting medium, Fluoromount™, Sigma-Aldrich®
Catalog # F4680-25ML
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Conf:25 mL
- Descrizione:Aqueous mounting medium
Specifications
About this item
Fluoromount™ Aqueous Mounting Medium for use with fluorescent dye-stained tissue.
Used for mounting:
- Coverslips on slides for immunofluorescence microscopic examination of PC12 and Hela cells
- Hippocampus neurons cells for confocal microscope studies
- Brain sections for immunofluorescence staining