Orders LinkContactUsLinkComponent
Differentiation solution, Sigma-Aldrich®
Differentiation solution, Sigma-Aldrich®
Differentiation Solution, Poly bottle, 1 L
Codice catalogo A3179-1L
Nome del fornitore:   SIGMA ALDRICH MICROSCOPY
Differentiation solution, Sigma-Aldrich®
Codice catalogo A3179-1L
Nome del fornitore:  SIGMA ALDRICH MICROSCOPY
Numero CAS:  
Differentiation solution, Sigma-Aldrich®
Differentiation Solution, Poly bottle, 1 L
Numero CAS:  
Price
SFr. 57.80
Log in per disponibilità
1 * 1 L
Product Details & Documents
Acid Alcohol. Acidified alcohol solution for the differentiation of regressive hematoxylin stains.
  • Suitable for Histology
  • Suitable for Hematology
Documents
Specifiche
Specifiche
  • Tipo di conf.:
    Poly bottle
  • Conf:
    1 L
  • Descrizione:
    Differentiation Solution
  • Cat. No.:
    A3179-1L
  • M-Clarity quality level:
    MQ200
  • N. MDL:
    MFCD00243254