Order Entry
Switzerland
ContactUsLinkComponent
Alluminio ossido neutro Brockmann Grade I 50-200 µm, 90A per cromatografia
Alluminio ossido neutro Brockmann Grade I 50-200 µm, 90A per cromatografia
Catalog # ACRO190410010
CAS Number:  
undefined
Alluminio ossido neutro Brockmann Grade I 50-200 µm, 90A per cromatografia
Catalog # ACRO190410010
CAS Number:  

Specifications

  • Tipo di conf.:
    Plastic bottle
  • Conf:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Particle size:
    ≤10 % (< 50 um)
  • Specific surface area:
    90 to 170 m2/g,N2 adsorp
  • pH:
    6.5 to 7.5

Specification Test Results

About this item

 :
 :101.96 g/mol
Boiling Pt:2980 °C (760 mmHg)
Melting Pt:2050 °C
Storage Temperature:Ambiente
CAS Number:1344-28-1
Formula:Al₂O₃
MDL Number:MFCD00003424