Spare LED tubes for Apollo II and Apollo II high intensity liquid viewers
Catalog # ADLM6031535
Supplier: ADELPHI MANUFACTURING
Specifications
- Descrizione:Apollo II LED tubes - T5
- Conf:2
Specifications
About this item
Pack of two 10 Watt LED bulbs (550 mm long) with a diffuser, which give a light level of between 2000 and 3750 Lux. The benefit of using LED tubes is more consistent light over the years compared with fluorescent tubes. These LED bulbs have a service life of 50,000 hours.
- Colour temperature: 4000 K
- Colour Rendering Index (CRI) Ra: greater than 80
- Luminous flux at 25 °C: 950 - 1150 Im
The same bulbs to be used for Apollo II (for transparent glass) as well as Apollo II High Intensity (for colour glass).