Hand Towels, KLEENEX® Ultra Slimroll™
KIMB6781
- Confezione:6 rolls/sheet, 2400 sheets/case
- Colore:bianco
- Descrizione:Kleenex® Ultra SLIMROLL™, hand towels, 2 ply
- L×L:100 m × 200 mm
- Conf:6 Roll
Kleenex® Ultra™ Slimroll™ is the most gentle rolled hand towel perfect for elevated care in premium washrooms. Slimroll™ hand towel pairs with our compact dispenser to deliver premium comfort and exceptional employee care.
- Ideal for environments where space may be limited
- Compatible with Aquarius™ Slimroll™ hand towel dispensers
The unique Airflex™ technology ensures that each lightweight single-ply towel is absorbent and strong, even when wet, preventing them from becoming soggy or ripping.