Orders LinkContactUsLinkComponent
CryoStorage vials
CryoStorage vials
Supplier:   EPPENDORF
CryoStorage vials
Supplier:  EPPENDORF
Certificates
CryoStorage vials
CryoStorage vials
10 Options Available Below
Product Details & Documents
CryoStorage vials for storage at −80 °C are labeled with a SafeCode barcode for safe sample identification. Different volumes are available.
  • Free of DNase, RNase, human DNA, endotoxin
  • External thread, pre-capped grey
  • With 2D SafeCode (DataMatrix/ECC200), linear and human readable code
Eppendorf CryoStorage vials take advantage of the 3-level coding of the SafeCode system (datamatrix code, barcode, and clear writing) to enable safe sample identification. Up to 96 pre-labelled vials are pre-racked and pre-capped for convenient usage. The vial range covers 0,5 to 4,0 ml for a broad range of storage applications at e.g. −80 °C in the ULT freezer. These vials are the solution for modern high-density storage with highest sample safety, management, and tracking needs.
Packaging: Individually packed racks.
Product Family Options
Product Information
  • Descrizione
    Capacità
    Tappo incluso
    Sterili
    Confezione
    Availability
    Price
  • CryoStorage vial
    0,50 ml
    Yes
    -
    10 racks of 96 vials
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - SFr. 798.00
    SFr. 798.00
    1 * 960 ST
       SFr. 798.00
  • CryoStorage vial
    1,00 ml
    Yes
    -
    10 racks of 96 vials
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - SFr. 829.00
    SFr. 829.00
    1 * 960 ST
       SFr. 829.00
  • CryoStorage vial
    1,50 ml
    Yes
    -
    10 racks of 48 vials
    Log in for availability
    Log in for availability
    1 * 480 ST
    1 * 480 ST - SFr. 548.00
    SFr. 548.00
    1 * 480 ST
       SFr. 548.00
  • CryoStorage vial
    2,00 ml
    Yes
    -
    10 racks of 48 vials
    Log in for availability
    Log in for availability
    1 * 480 ST
    1 * 480 ST - SFr. 564.00
    SFr. 564.00
    1 * 480 ST
       SFr. 564.00
  • CryoStorage vial
    4,00 ml
    Yes
    -
    10 racks of 48 vials
    Log in for availability
    Log in for availability
    1 * 480 ST
    1 * 480 ST - SFr. 733.00
    SFr. 733.00
    1 * 480 ST
       SFr. 733.00
  • CryoStorage vial
    0,50 ml
    Yes
    +
    10 racks of 96 vials
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - SFr. 850.00
    SFr. 850.00
    1 * 960 ST
       SFr. 850.00
  • CryoStorage vial
    1,00 ml
    Yes
    +
    10 racks of 96 vials
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - SFr. 1’640.00
    SFr. 1’640.00
    1 * 960 ST
       SFr. 1’640.00
  • CryoStorage vial
    1,50 ml
    Yes
    +
    10 racks of 48 vials
    Log in for availability
    Log in for availability
    1 * 480 ST
    1 * 480 ST - SFr. 574.00
    SFr. 574.00
    1 * 480 ST
       SFr. 574.00
  • CryoStorage vial
    2,00 ml
    Yes
    +
    10 racks of 48 vials
    Log in for availability
    Log in for availability
    1 * 480 ST
    1 * 480 ST - SFr. 590.00
    SFr. 590.00
    1 * 480 ST
       SFr. 590.00
  • CryoStorage vial
    4,00 ml
    Yes
    +
    10 racks of 48 vials
    Log in for availability
    Log in for availability
    1 * 480 ST
    1 * 480 ST - SFr. 760.00
    SFr. 760.00
    1 * 480 ST
       SFr. 760.00