Orders LinkContactUsLinkComponent
Restroom bundle with adhesive backing, PIG®
Restroom bundle with adhesive backing, PIG®
Restroom bundle, 5 pads per box, 0,8 kg, Polypropylene, 480×610 mm
Codice catalogo 115-2579
Nome del fornitore:   NEW PIG
Restroom bundle with adhesive backing, PIG®
Codice catalogo 115-2579
Nome del fornitore:  NEW PIG
Restroom bundle with adhesive backing, PIG®
Restroom bundle with adhesive backing, PIG®
Restroom bundle, 5 pads per box, 0,8 kg, Polypropylene, 480×610 mm
Price
SFr. 76.70
Log in per disponibilità
1 * 5 ST
Product Details & Documents
Adhesive-backed mat bundle has everything you need to keep the floor in a small restroom clean, dry and safe, Includes urinal, toilet, plunger, and sink and dryer mats.
  • Bundle includes two sizes of urinal mats, toilet mat, sink/dryer mat and plunger mat for maximum drip/splash coverage
  • Permanently infused antimicrobial technology prevents the growth of odor-causing bacteria and mold on the mats to keep restrooms cleaner and fresher smelling
  • Proprietary, full-coverage adhesive backing keeps mats in place underfoot with no shifting, curling or flipping up to stop slip and trip hazards; peels up easily
  • Mats stick to your floor and won't budge while you clean them; simply mop them with the rest of the floor
  • Durable, Hi-Trac™ surface stands up to daily cleaning and foot traffic
  • Absorbs water and urine
EFM™ Absorbent technology captures water on contact, wicks it away and dries out quickly, impermeable backing layers stop liquid pass-through to prevent stains and grout damage.
Release paper on the bottom of the mat should be removed prior to installation.
Certifications: Tested and certified by the National Floor Safety Institute (NFSI) as a high-traction surface which can reduce slip and fall claims by as much as 90% when used as part of a floor safety program.
Attenzione: Mats take on the properties of absorbed liquids - always take proper precautions when handling saturated absorbents.
Specifiche
  • Tipo:
    Polypropylene
  • Descrizione:
    Restroom bundle, 5 pads per box, 0,8 kg
  • L×L:
    480×610 mm