Auftragserfassung
Switzerland
ContactUsLinkComponent
Poly CSXB-35, J.T. Baker® BAKERBOND® XWP 500
Poly CSXB-35, J.T. Baker® BAKERBOND® XWP 500
  7572-01
 :  Avantor
 :  
undefined
Poly CSXB-35, J.T. Baker® BAKERBOND® XWP 500
  7572-01
 :  Avantor
 :  
  • Dimensioni particelle:
    35 µm
  • Conf:
    50 mL
  • Dimensioni pori:
    500 Å
  • Descrizione:
    Poly CSxB (hydrophilic polymer-CSX), weak anion exchanger media
  • Seq:
    08
  • pH di esercizio:
    2-14
Polimero CSX idrofilico.
  • Conservare tra 2 e 15 °C