Nitrite test strips, Dosatest
Nitrite test strips, 0 • 0.1 • 0.3 • 0.6 • 1 • 2 • 3 g/L NO₂⁻, Griess' reaction, 100, 100
Nome del fornitore: VWR Chemicals
Nitrite test strips, Dosatest
Codice catalogo 85705.601
Nome del fornitore: VWR Chemicals
Numero CAS:
Vai a
Product Details & Documents
Semi-quantitative test strips for the determination of nitrite.
- Ideal for rapid and easy analysis of nitrite directly on-site
- No maintenance or calibration is required
Colour change: Yellow to red
Informazioni per l'ordine: Delivered as 100 test strips in a tube, instruction leaflet in a box
Attenzione: Strongly acidic solutions must be diluted with dilute sodium hydroxide and strongly alkaline samples with dilute sulphuric acid to pH 4 to 6.
Documents
Certificates
Cerca per numero di lottoSDS_IT_CH_VWRC85705 sds_VWRC85705
Scheda di sicurezza
VWR Indicator Papers and Strips.pdf
Letteratura sul prodotto
Specifiche
Specifiche
- Metodo chimico:Griess' reaction
- Conf:100
- Descrizione:Nitrite test strips
- Test per kit:100
- Range di misura:0 • 0.1 • 0.3 • 0.6 • 1 • 2 • 3 g/L NO₂⁻