Ascorbic acid test strips, Dosatest
Ascorbic acid test strips, 0 • 50 • 100 • 200 • 300 • 500 • 700 • 1000 • 2000 mg/L Vitamin C, Phosphormolybdenum blue, 100, 100
Nome del fornitore: VWR Chemicals
Ascorbic acid test strips, Dosatest
Codice catalogo 85701.601
Nome del fornitore: VWR Chemicals
Numero CAS:
Vai a
Product Details & Documents
Semi-quantitative test strips for the determination of ascorbic acid.
- Ideal for rapid and easy analysis directly on-site
- No maintenance or calibration is required
Colour change: Yellow to green-blue
: Delivered as 100 test strips in a tube with instruction leaflet.
Specifiche
Specifiche
- Metodo chimico:Phosphormolybdenum blue
- Conf:100
- Descrizione:Ascorbic acid test strips
- Test per kit:100
- Range di misura:0 • 50 • 100 • 200 • 300 • 500 • 700 • 1000 • 2000 mg/L Vitamin C