D-Ring attachment with cord, DBI-SALA, 3M™
Catalog # CPSD1500009
Supplier: CAPITAL SAFETY
Specifications
- Descrizione:D-Ring attachment with cord
- Conf:10
Specifications
About this item
D-Ring cord attachment point, 2,3 kg (5 lbs.) capacity.
- Adjusts to virtually any wrist size with hook and loop system
- Replaceable steel cable retractor fits into a sewn-in pocket and allows tools to easily be tied off using an attached thumb-operated trigger
- Elastic band helps ensure a comfortable fit
- Attached trigger can be used to quickly work with many hand tools
The DBI SALA® D-ring cord easily cinches to tools weighing under 2,3 kg creating an instant attachment point for many tools.
Designed for use in general industrial, oil and gas, mining, construction, and transportation industries.