ContactUsLinkComponent
High-temperature silicone cap liner, GL 45, PTFE coated, DURAN®
High-temperature silicone cap liner, GL 45, PTFE coated, DURAN®
  215-4281
   Riduzione degli sprechi
undefined
High-temperature silicone cap liner, GL 45, PTFE coated, DURAN®
  215-4281
   Riduzione degli sprechi
  • Descrizione:
    High-temperature cap liner, GL 45, PTFE coated FKM, 43,5×3,0 mm
  • Environmentally Preferable:
    Riduzione degli sprechi
  • Conf:
    10
Replacement GL 45 PTFE coated high-temperature silicone cap liner for the aluminium cap.