Dual auto-reader for steam and vaporised hydrogen peroxide 490, Attest™
Catalog # MMME490
Supplier: 3M MEDICAL
Specifications
- Descrizione:Dual auto-reader for steam and vaporised hydrogen peroxide 490, model 1496VF
- Conf:1
Specifications
About this item
Auto-reader for use with 3M™ Attest™ Rapid Readout Biological Indicators and process challenge packs for steam. Includes ethernet cable for web app connectivity.
- Designed for use with Attest™ Super Rapid Readout Biological Indicators 1491 and 1492
- Intelligent display with time-to-result countdown feature
- Display viewable remotely using your Smartphone or PC, manage record-keeping with web app and your internet connected PC
- Reader will still connect to leading instrument Tracking Software Systems
The 3M™ Attest™ 490 Auto reader for steam reads the fluorescence produced by positive 3M™ Attest™ Rapid Readout Biological Indicators 1491 and 1492 to indicate a sterilisation process failure. Featuring advanced electronics and backlit LCD panel, the 3M™ Attest™ Auto reader 490 turns up the dial on reliability and accuracy. It gives you even more confidence in your efforts to ensure patient safety, while working with your existing 3M™ Attest™ Rapid Readout products.
Test substance: Hydrogen peroxide