Earmuffs, WS™ LiteCom Pro III Headset, Peltor™
Product Details & Documents
The 3M™ Peltor™ WS™ LiteCom Pro III Headset features an integrated, programmable analog/digital two-way radio, Bluetooth® Multipoint connectivity, noise-cancelling microphone, and level-dependent function for ambient listening. The headset not only helps protect against hazardous noise, but also makes it easy to communicate in noisy environments and maintain situational awareness.
- Programmable analog/digital DMR two-way radio communication
- Bluetooth® multipoint functionality
- Water tight microphone (IP 68)
- Level-dependent function for ambient listening
- Voice menu with multiple language selection
- Push-to-Listen function
- Rechargeable battery, Li-Ion
- Voice operated transmission (VOX)
The Peltor™ WS™ LiteCom Pro III Headset offers wireless communication in noisy environments. Suitable for work in industrial and construction applications, where hands-free communication and hearing protection are needed. Additional features include busy channel indication, an external connection jack, auto power off function, and battery status indication.
Certifications: Conforms with the provisions set out in the PPE directive 89/686/EEC and RED directive 2014/53/EU.
Specifications
- Colore:nero/blu
- Descrizione:Peltor™ WS™ LiteCom Pro III Headset, 403-470 MHz, analog/digital, EU version, headband