General purpose gloves, High performance, A700
Informazioni su questo articolo
This high-performing, hand-hugging glove is multi-purpose and suitable for any job.
- Reinforced stitching on the thumb for extra strength
- Low linting construction for minimal contamination
- Wrist support and protection
- Wrist strap for secure fitting
Lightweight for the wearer, this glove has durability built in and features reinforced stitching on the thumb for extra strength with wrist support and protection.
Suitable for mechanic applications, general construction, material handling, carpentry and utility work.
Certifications: EN 388:2016 (1111X)
4 varianti
Product Details & Documents
This high-performing, hand-hugging glove is multi-purpose and suitable for any job.
- Reinforced stitching on the thumb for extra strength
- Low linting construction for minimal contamination
- Wrist support and protection
- Wrist strap for secure fitting
Lightweight for the wearer, this glove has durability built in and features reinforced stitching on the thumb for extra strength with wrist support and protection.
Suitable for mechanic applications, general construction, material handling, carpentry and utility work.
Certifications: EN 388:2016 (1111X)
Specifications for Product Family
Specifiche
- Catalog No:
- PRTWA700BKRM
- PRTWA700BKRL
- PRTWA700BKRXL
- PRTWA700BKRXXL
- Modello:
- A700
- A700
- A700
- A700
- Ambidestro/mano specifico:
- mano specifico
- mano specifico
- mano specifico
- mano specifico
- Monouso/riutilizzabile:
- riutilizzabile
- riutilizzabile
- riutilizzabile
- riutilizzabile
- Tipo polsino:
- Fascetta regolabile in Velcro
- Fascetta regolabile in Velcro
- Fascetta regolabile in Velcro
- Fascetta regolabile in Velcro
- Norma EN:
- EN 388:2016 (1111X)
- EN 388:2016 (1111X)
- EN 388:2016 (1111X)
- EN 388:2016 (1111X)
Specifiche
Product Family Options
Informazioni sul prodotto
- Dimensione del guantoColoreDisponibilitàPrice
- Specifiche:
More
Specifications
Cat. No.PRTWA700BKRMModelloA700Ambidestro/mano specificomano specificoMonouso/riutilizzabileriutilizzabileTipo polsinoFascetta regolabile in VelcroNorma ENEN 388:2016 (1111X) - Specifiche:
More
Specifications
Cat. No.PRTWA700BKRLModelloA700Ambidestro/mano specificomano specificoMonouso/riutilizzabileriutilizzabileTipo polsinoFascetta regolabile in VelcroNorma ENEN 388:2016 (1111X) - Specifiche:
More
Specifications
Cat. No.PRTWA700BKRXLModelloA700Ambidestro/mano specificomano specificoMonouso/riutilizzabileriutilizzabileTipo polsinoFascetta regolabile in VelcroNorma ENEN 388:2016 (1111X) - Specifiche:
More
Specifications
Cat. No.PRTWA700BKRXXLModelloA700Ambidestro/mano specificomano specificoMonouso/riutilizzabileriutilizzabileTipo polsinoFascetta regolabile in VelcroNorma ENEN 388:2016 (1111X)