Orders LinkContactUsLinkComponent
Cork stoppers assortment
Cork stoppers assortment
Codice catalogo 217-9654
Nome del fornitore:   BRAEMSWIG
Cork stoppers assortment
Codice catalogo 217-9654
Nome del fornitore:  BRAEMSWIG
Cork stoppers assortment
Cork stoppers assortment
Price
69.30 SFr.
Log in per disponibilità
1 * 100 ST
Product Details & Documents
Natural cork, A1 quality
  • Lightweight, tough
  • Easy to drill holes into
  • Very economical
The assortment 217-9654 contains 10 stoppers each of Cat No. 217-1012/-1014/-1016/-1017/-1018/-1019/-1020/-1021/-1022 and 217-1024.