Spill control pads
Spill control pads, PP, grey, 0,8 L
Codice catalogo PRTWSM20GRR
Nome del fornitore: Portwest
Vai a
Product Details & Documents
These pads have open edges for quick wicking. High strength spunbound coverstocks prevent lint sticking to machine parts and hold the pads together even when saturated.
- Each pad absorbs 0,5 litres
Certifications: BS 7959-1, BS 7959-3
Specifiche
- Descrizione:Spill control pads, PP, grey
- Assorbanza:0,8 L