Welding helmet, BizWeld Plus, PW65
PRTWPW65BKR
: Portwest
- Descrizione:Welding helmet, black
- Materiale:PP/PE
- Modello:PW65
- Conf:1
Auto-darkening welding helmet, which switches from a light to dark state when welding activity starts.
- Wheel ratchet size adjustment for easy fitting
- Solar cell power with two built-in lithium back-up batteries and a working time of over 5000 hours
- Auto-darkening, switches from light to dark when the welding process starts
- Suitable for a wide range of applications
- Suitable to be worn with disposable respirators
- Retail box which aids presentation for retail sales
Response time from light to dark state is 0,04 milliseconds; return from dark to clear state in 0,03 to 0,04 second.
Power is supplied by solar cells with 2 built-in lithium batteries to offer power back-up for longer service life. No need for replacement batteries.
Standard: EN 166 1 S, EN 175 F, EN 379 4 / 9-13 ; 1/1/1/2.