Gas filters, P926
Catalog # PRTWP926BKR
Supplier: Portwest
Specifications
- Descrizione:ABEK2 Gas filter, P906
- Conf:4
Specifications
About this item
P926 filters are anti-gas and protects against contaminant gases. Equipped with a standard threaded connection conforming to EN 148-1 and can be used on full face masks equipped with the same standard connection.
- Lightweight and comfortable
- Individually packed for vending machines
These class 2 filters protects from organic and inorganic acid gases and vapours, sulphur dioxide, ammonia and its derivates where the boiling point is higher than 65 °C.
Certifications: EN 148 -1, EN 14387 Type ABEK Class 2