Filter for half masks, P940
PRTWP940GRR
: Portwest
- Tipo:Particle filter, P940, grey
- Conf:6
- Classe:P3
P940 is a particle filter that protects against dusts. It is equipped with a special thread for pair use only and it can be used only on the P410 half mask.
- Lightweight and comfortable
- Screw-in connection
This filter is a P3 R type and protects against toxic substances such as particles and microorganisms.
: EN 143 Type P 3R