Rain jackets, S571
Informazioni su questo articolo
A classic jacket with good fit to streamline and reduce the bulkiness women often feel in a fourseason jacket. The longer line back panel and discreet waist adjustment make this a contemporary silhouette and give a flattering smooth front jacket.
- 3 in 1 waterproof and windproof jacket
- Extremely water resistant fabric finish, water beads away from fabric surface
- Taped seams to provide additional protection
- Interactive inner garment
- Interactive fleece which can be worn as a separate jacket for multi use
- Five pockets for ample storage which are zipped phone pocket
- Two-way zip for quick and easy access
- Hook and loop closure for easy access
The following Rainwear collections offer an outstanding choice of designs and colours, suitable for many end users. The best quality fabrics and high technology construction techniques are used throughout, guaranteeing the ultimate in all-weather protection. Tested to withstand the rigours of everyday wear, each garment has been designed and manufactured with care to ensure total satisfaction wear after wear.
100% polyester, Pongee PVC coated shell fabric, 100% polyester lining fabric with 100% polyester fleece removable fabric.
Certifications: EN 343 Class 3:1 (WP 5,000 mm).
2 varianti
Product Details & Documents
A classic jacket with good fit to streamline and reduce the bulkiness women often feel in a fourseason jacket. The longer line back panel and discreet waist adjustment make this a contemporary silhouette and give a flattering smooth front jacket.
- 3 in 1 waterproof and windproof jacket
- Extremely water resistant fabric finish, water beads away from fabric surface
- Taped seams to provide additional protection
- Interactive inner garment
- Interactive fleece which can be worn as a separate jacket for multi use
- Five pockets for ample storage which are zipped phone pocket
- Two-way zip for quick and easy access
- Hook and loop closure for easy access
The following Rainwear collections offer an outstanding choice of designs and colours, suitable for many end users. The best quality fabrics and high technology construction techniques are used throughout, guaranteeing the ultimate in all-weather protection. Tested to withstand the rigours of everyday wear, each garment has been designed and manufactured with care to ensure total satisfaction wear after wear.
100% polyester, Pongee PVC coated shell fabric, 100% polyester lining fabric with 100% polyester fleece removable fabric.
Certifications: EN 343 Class 3:1 (WP 5,000 mm).
Specifications for Product Family
Specifiche
- Catalog No:
- PRTWS571NARXS
- PRTWS571NARXL
- Modello:
- S571
- S571
- Applicazioni per abiti:
- General Purpose
- General Purpose
- Risk area:
- Rain (waterproof)
- Rain (waterproof)
- Genere:
- Per le donne
- Per le donne
- Sleeve or leg length:
- Long
- Long
- Tipo di chiusura:
- Con chiusura a zip
- Con chiusura a zip
- Tessuto:
- 100% polyester
- 100% polyester
- Peso:
- 550 g
- 550 g
Specifiche
Product Family Options
Informazioni sul prodotto
- ColoreTaglia dell'abbigliamentoDisponibilitàPrice
- Specifiche:
More
Specifications
Cat. No.PRTWS571NARXSModelloS571Applicazioni per abitiGeneral PurposeRisk areaRain (waterproof)GenerePer le donneSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessuto100% polyesterPeso550 g - Specifiche:
More
Specifications
Cat. No.PRTWS571NARXLModelloS571Applicazioni per abitiGeneral PurposeRisk areaRain (waterproof)GenerePer le donneSleeve or leg lengthLongTipo di chiusuraCon chiusura a zipTessuto100% polyesterPeso550 g