Work waistcoats, Iona Executive, F476
Supplier: Portwest
Certificates
About this item
This superior vest option offers a Dual ID window, radio loop and multi-functional pockets as standard. Perfect for corporate branding.
- Reflective tape for increased visibility
- Front zip opening for easy access
- Two lower patch pockets
- Phone and pen holder pocket
- Radio loop for easy clipping of a radio
- 7 pockets for ample storage
- Retail bag which aids presentation for retail sales
Features D-ring for keys or ID cards and dual ID holder allows use of both portrait and landscape ID card display.
Shell fabric: 100% polyester, warp knit, 125 g
Certifications: AS 4602.1 Class N
Specifications
- Catalog No:
- PRTWF476NARXL
- PRTWF476NARS
- PRTWF476NARM
- PRTWF476NARL
- PRTWF476NARXXL
- PRTWF476RBRL
- PRTWF476RBRS
- PRTWF476RBRXL
- PRTWF476RERM
- PRTWF476RERXXXL
- PRTWF476NARXXXL
- PRTWF476BGRL
- PRTWF476BGRXL
- PRTWF476BGRXXL
- PRTWF476BGRS
- PRTWF476BGRXXXL
- PRTWF476RERXL
- PRTWF476BGRM
- PRTWF476RERS
- PRTWF476RBRXXL
- PRTWF476RBRM
- PRTWF476RERL
- PRTWF476RBRXXXL
- PRTWF476RERXXL
- Modello:
- F476
- F476
- F476
- F476
- F476
- F476
- F476
- F476
- F476
- F476
- F476
- F476
- F476
- F476
- F476
- F476
- F476
- F476
- F476
- F476
- F476
- F476
- F476
- F476
- Applicazioni per abiti:
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- Risk area:
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- Genere:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Tipo di chiusura:
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Con chiusura a zip
- Tessuto:
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- Peso:
- 125 g
- 125 g
- 125 g
- 125 g
- 125 g
- 125 g
- 125 g
- 125 g
- 125 g
- 125 g
- 125 g
- 125 g
- 125 g
- 125 g
- 125 g
- 125 g
- 125 g
- 125 g
- 125 g
- 125 g
- 125 g
- 125 g
- 125 g
- 125 g