High visibility rain trousers, H441
Supplier: Portwest
Certificates
About this item
High visibility rain trousers are lightweight and extremely practical. They can be layered over the wearers own trousers for complete waterproof protection.
- Reflective tape for increased visibility
- Stud adjustable hems for a secure fit
- Twin-stitched seams for extra durability
- 50+ UPF rated fabric to block 98% of UV rays
- Wind resistant to protect against wind chill
Shell fabric : 100% polyester, 190T, plain weave, PVC coated 200 g.
Extremely water resistant fabric finish, water beads away from fabric surface. Fully elasticated waistband for ultimate wearer comfort.
Certifications: Standards: ANSI/ISEA 107: 2015 SUPPLEMENTAL CLASS E, AS/NZS 1906.4, EN 343:2019 Class 3:1 X WP 5,000mm, EN ISO 20471 Class 1.
Caution: Wash with like colours. Do not dry clean.
Specifications
- Catalog No:
- PRTWH441ORRM
- PRTWH441ORRXXXL
- PRTWH441ORRS
- PRTWH441ORRXL
- PRTWH441YERXXL
- PRTWH441YERXL
- PRTWH441ORRL
- PRTWH441ORRXXL
- PRTWH441YERL
- PRTWH441YERM
- PRTWH441YERS
- PRTWH441YER4XL
- PRTWH441YERXXXL
- PRTWH441YERXS
- Modello:
- H441
- H441
- H441
- H441
- H441
- H441
- H441
- H441
- H441
- H441
- H441
- H441
- H441
- H441
- Risk area:
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- Genere:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Tessuto:
- 100% polyester/PVC coated
- 100% polyester/PVC coated
- 100% polyester/PVC coated
- 100% polyester/PVC coated
- 100% polyester/PVC coated
- 100% polyester/PVC coated
- 100% polyester/PVC coated
- 100% polyester/PVC coated
- 100% polyester/PVC coated
- 100% polyester/PVC coated
- 100% polyester/PVC coated
- 100% polyester/PVC coated
- 100% polyester/PVC coated
- 100% polyester/PVC coated
- Peso:
- 200 g/m²
- 200 g/m²
- 200 g/m²
- 200 g/m²
- 200 g/m²
- 200 g/m²
- 200 g/m²
- 200 g/m²
- 200 g/m²
- 200 g/m²
- 200 g/m²
- 200 g/m²
- 200 g/m²
- 200 g/m²