Earplugs, PU foam, corded, EP08
Catalog # PRTWEP08ORR
Supplier: Portwest
Specifications
- Colore:arancione
- Descrizione:Ear plugs, corded
- Conf:200 PAIR
Specifications
About this item
These disposable ear plugs are made from soft PU foam and PVC.
- Corded for safe keeping
- Each pair inside individual hygienic polybag
- Excellent attenuation rate
- Retail box which aids presentation for retail sales
- Individually packed for vending machines
These are designed with latest standards to deliver the highest level of protection. Comfortable and lightweight and ensure a pleasant wear, even for long periods of use.
Certifications: EN 352-2 (SNR 36dB); ANSI S3.19 (32dB).
Packaging: 200/pack, 10/carton.