ContactUsLinkComponent
Leather gloves, classic driver, A225, Insulatex™
  PRTWA225RERXL
 :  Portwest
undefined
Leather gloves, classic driver, A225, Insulatex™
  PRTWA225RERXL
 :  Portwest
  • Dimensione del guanto:
    XL
  • Ambidestro/mano specifico:
    mano specifico
  • Colore:
    rosso/yellow
  • Modello:
    A225
  • Conf:
    1 PAIR
Insulated leather rigger with fleece lining for added warmth when working in cold environments.
  • Specially designed for use in cold conditions
  • Superb abrasion and tear resistance
  • Classic leather driver
Materials: cow split leather, Insulatex™.
 : Standards: AS/NZS 2161 .2, AS/NZS 2161 .3, AS/NZS 2161 .5, EN388:2016 +A1:2018 - (4.2.4.2.X), EN 511 (1.1.X), EN 420.