Wrap around glasses, PW32
PRTWPW32CLR
: Portwest
- Tipo:UV protection, anti scratch
- Colore:nero
- Materiale lente:Clear PC
- Conf:1
Wrap-around design glasses with one piece lens.
- 99% UV protection
- Anti-scratch coating for added durability
- Flexible arms for ultra comfort
- Ultra lightweight for long uses
- Individually packed for vending machines
- Retail bag which aids presentation for retail sales
: AS/NZS 1337.1; EN 170 (2C-1.2 (clear)); ANSI/ISEA Z87 + (U6 (Clear))
: Cord included.