Flexible fit probed test earplugs, E-A-R™
MMMA393-2026-50
: 3M
- Descrizione:Flexible fit probed test earplugs
- Conf:50 PAIR
E-A-R™ Flexible Fit Earplugs equipped with a probe tube for fit testing using the 3M™ E-A-Rfit™ Dual-Ear Validation System. Cannot be used for hearing protection.
- Test earplugs are modified with a probe tube for use during earplug fit testing
- 50 pairs per case
- For occupational use only
These test Plugs are a specially probed version of the standard earplug. This product works with the 3M™ E-A-Rfit™ Dual-Ear Validation System to allow fit tests of Flexible Fit earplugs.