Darkfield optic ring for VisiLED slim ring light
Darkfield optic ring for VisiLED slim ring light
Catalog # SCOC400810
Supplier: SCHOTT AG Lighting and imaging
Darkfield optic ring for VisiLED slim ring light
Catalog # SCOC400810
Supplier: SCHOTT AG Lighting and imaging
Jump to:
Product Details & Documents
Focus optic ring for darkfield illumination.
- Focus optics ring for darkfield
- Working distance: 5 to 25 mm
Designed for use only with VisiLED slim ring light (SCOC400150).
Ordering information: VisiLED slim ring light and controller MC 750, MC 1100, or MC 1500 required.
Documents
TDS VisiLED Slim Ringlight
Datasheet
Specifications
- Descrizione:Darkfield optic ring for VisiLED slim ring light