Polariser set for ring light, VisiLED, S40-75
Catalog # SCOC400575
Supplier: SCHOTT AG Lighting and imaging
Specifications
- Descrizione:Polariser filter set for ring light S40-75
- Environmentally Preferable:
- Conf:1
Specifications
About this item
Polarisation system for brightfield ringlights. Turnable polariser with adapting thread and sliding filter as analyser.
- Polarised light by a polarising and analysing filter within the beam path can be very efficiently used in stereo microscopy
- Use to contrast inner material structure or to identify metal particles within others
Designed for use with VisiLED ring light S40-75.
Ordering information: VisiLED ringlight S40-75 and controller MC 750, MC 1100, or MC 1500 required.