Molybdenum ICP and ICP-MS standards, (CRMs), Fluka™
Catalog # HONCE1401-100ML
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Concentrazione:10 mg/L
- Matrice:H₂O
- Tipo di conf.:Plastic bottle
- Conf:100 mL
- Componenti:Molibdeno
- Simbolo:Mo
Specifications
About this item
Certified Reference Materials (CRMs)
- Traceable to SRM from NIST
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).