Iron ICP and ICP-MS Standards, (CRMs), Fluka™
HONCE1386-100ML
:
- Concentrazione:10 mg/L
- Matrice:2% HNO₃
- Tipo di conf.:Plastic bottle
- Conf:100 mL
- Componenti:Ferro
- Simbolo:Fe
Certified Reference Materials (CRMs)
- Traceable to SRM from NIST
: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).