About this item
The Panthera TEC POL models present a high flexibility for the inspection of all kind of polarizing materials, for transparent samples like fibers and foils with their inherent birefringence, as well as for opaque specimen from education environment and industrial quality control.
- CCIS® UC Plan Achromat, DIN objectives (strain-free) for FOV 22 mm
- Transmitted and transmitted/incident stand options
- Transmitted light: LEDs/HAL light source interchangeable
- Integrated focusable/centerable Bertrand lens
- Motic LightTracer: coded nosepiece and digital light intensity knob
- Packaging size: 630×370×590 mm
- Power supply 110 to 240 V
These microscopes are equipped with Bertrand lens to analyse the crystal structure and interference figures of gems and minerals. Choose your model between transmitted light only, or transmitted and epi/incident light with or without integrated camera.
Even transparent birefringent structures from biology and medicine (heart muscle cells, secondary cell walls, and so on) can be detected by the Full Koehler illumination with exchangeable LEDs/halogen light source. In Incident light, a 3 W LED is integrated in a Brightfield illumination setup with Aperture- and Field diaphragm. The Panthera TEC POL models feature UC Plan Achromat objectives with focus on a strain-free mounting for maximum darkness of the image background.
Specifications
- Catalog No:
- 630-3120
- 630-3119
- 630-3121
- 630-3118
- 630-3124
- Modello:
- Panthera TEC POL Epi Binocular
- Panthera TEC POL Trinocular
- Panthera TEC POL Epi Trinocular
- Panthera TEC POL Binocular
- Panthera TEC POL (i) Digital with integrated WiFi camera
- Metodo d’osservazione:
- Campo chiaro e polarizzazione
- Campo chiaro e polarizzazione
- Campo chiaro e polarizzazione
- Campo chiaro e polarizzazione
- Campo chiaro e polarizzazione
- Testa:
- Inclinazione a 25°, rotazione a 360°
- Inclinazione a 25°, rotazione a 360°
- Inclinazione a 25°, rotazione a 360°
- Inclinazione a 25°, rotazione a 360°
- Inclinazione a 25°, rotazione a 360°
- Oculari:
- UC-WF 10×/22 mm
- UC-WF 10×/22 mm
- UC-WF 10×/22 mm
- UC-WF 10×/22 mm
- UC-WF 10×/22 mm
- Revolver portaobiettivi:
- 5 posizioni, invertito
- 5 posizioni, invertito
- 5 posizioni, invertito
- 5 posizioni, invertito
- 5 posizioni, invertito
- Obiettivi:
- Acromatico 4×, 10×, 40×, 60×
- Acromatico 4×, 10×, 40×, 60×
- Acromatico 4×, 10×, 40×, 60×
- Acromatico 4×, 10×, 40×, 60×
- Acromatico 4×, 10×, 40×, 60×
- Tavolino:
- Circular rotating 360°, lockable, Ø 160 mm, 1° increments, 0,1° vernier scale
- Circular rotating 360°, lockable, Ø 160 mm, 1° increments, 0,1° vernier scale
- Circular rotating 360°, lockable, Ø 160 mm, 1° increments, 0,1° vernier scale
- Circular rotating 360°, lockable, Ø 160 mm, 1° increments, 0,1° vernier scale
- Circular rotating 360°, lockable, Ø 160 mm, 1° increments, 0,1° vernier scale
- Messa a fuoco:
- Coaxial coarse and fine focusing system with tension adjustment
- Coaxial coarse and fine focusing system with tension adjustment
- Coaxial coarse and fine focusing system with tension adjustment
- Coaxial coarse and fine focusing system with tension adjustment
- Coaxial coarse and fine focusing system with tension adjustment
- Fotocamera:
- None
- None
- None
- None
- Integrated camera with chip ¹/₃", data transfer: Wi-Fi, ethernet. Capture resolution 4 MP (2592×1520), live display mode via WiFi or ethernet 1280×720, 1920×1080 (full HD)
- Peso:
- 15,4 kg
- 15,4 kg
- 15,4 kg
- 15,4 kg
- 15,4 kg