Centrifugal filtration units, Vivaspin® Turbo 15 RC
Catalog # 512-0753
Supplier: Sartorius
Specifications
- Membrana:Regenerated cellulose
- MWCO:10 kD
- Seq:03
- Conf:12
- Volume del campione:4 - 15 ml
Specifications
About this item
Vivaspin® Turbo 15 Regenerated Cellulose (RC) membrane, ultrafiltration centrifugal concentrators for protein, DNA, nanoparticle and virus concentration for 4 to 15 ml sample volumes.
- Unique optimal membrane choice (PES and RC)
- Complete recoveries
- Fastest concentrations
- Highest chemical resistance
- Easy retentate collection
- Graduated and markable
Vivaspin® Turbo centrifugal concentrators offer the optimal choice for maximum concentration or buffer exchange applications, with broad range of MWCOs.
The Vivaspin® Turbo 15 RC concentrators are the Regenerated Cellulose concentrators with the highest flow rates on the market due to large, twin vertical membranes and sleek internal profile, which minimise protein polarization and subsequent fouling of the membrane and ensures maximum process speeds right down to the last 100 µl.