X-ray and metal detectable pallet knife and stirrer
Codice catalogo SAMPM1016B-200
Nome del fornitore: Sampling Systems
Specifiche
- Sterili:-
- Conf:4
- Lunghezza lama:200 mm
Specifiche
Informazioni su questo articolo
The X-ray and metal detectable pallet knife is ideal for scraping and mixing material. The pallet knives are made from a special grade of PP that can be detected. The bright blue colour also means that they can be easily seen if left in the material in line with HACCP guidelines.
- Robust PP construction, provides excellent chemical resistance to many products
- Long blade, can be used to stir and mix containers
- Full batch traceability
- The 200 mm blade can be used to stir and mix containers
To ensure the cleanliness of the pallet knives are manufactured and packed in a cleanroom. They can be used as a single use disposable product straight from the bag or reused.
Certifications: These PP pallet knives conform to all the latest standards including FDA 177.1520, EU 10/2011, EC 1935/2004 and are BSE/TSE free. Batch certificates are available.
Packaging: Supplied individually bagged.